Non Irradiated Contact Plates Media, BioMerieux
Supplier: bioMerieux
For detection and enumeration of microorganisms present on surfaces of sanitary importance.
Tryptic Soy Agar with Lecithin and Polysorbate 80 is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments.
D/E agar is a medium used for the detection and enumeration of microorganisms that are present in environments that contain antimicrobial chemicals, such as surfaces that are of sanitary importance.
Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
Ordering information: Made to order Items have a lead time of 42 days.