Irradiated 150 mm Plates Media, BioMerieux
Supplier: bioMerieux
L2037-TI
L2037-IR
95061-262PK
108.89
USD
95061-262
95061-260
Irradiated 150 mm Plates Media, BioMerieux
Media for Microbiology
Prepared Media for Microbiology
These contact plates can be used for surface environmental monitoring.
Tryptic Soy Agar is a general-purpose solid medium used for the cultivation and enumeration of a wide variety of fastidious and non-fastidious micro-organisms. Also known as Soybean-Casein Digest Agar (USP).