Irradiated 100 mm Plates Media, BioMerieux
Supplier: bioMerieux
43711 M1025-IR M1025-TI M1060-IR M1219-IR 43287 M1040-IR
95060-810PK 568.05 USD
95060-810 95060-832 95061-318 95061-324 89131-936 95061-294 95061-298 95061-330 95061-398 95061-334 95060-814 95061-314 95061-410
Irradiated 100 mm Plates Media, BioMerieux
Media for Microbiology Prepared Media for Microbiology
These contact plates can be used for surface environmental monitoring.
Sabouraud Dextrose Agar is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
Irradiated Tryptic Soy Agar (TSA) is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments also used for viable air monitoring.
Irradiated Dey-Engley Neutralizing Agar is used for the detection and enumeration of micro-organisms that are present in environments that contain antimicrobial chemical, such as surfaces that are of sanitary importance.
Irradiated Tryptic Soy Agar (TSA) with Lecithin and Polysorbate 80 is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments.
Trypcase soy 3P™ agar with neutralizers and lactamase is intended for the monitoring of microbial contamination in pharmaceutical industry cleanrooms.
Ordering information: Made to order items have a lead time of 42 days.