Starch Agar
Supplier: VWR International
470317-442
470317-444
470317-442EA
61.85
USD
470317-442
470317-444
Starch Agar
Media for Microbiology
Prepared Media for Microbiology
Used to determine if bacteria produce extracellular enzymes to catalyze the hydrolysis of starch. Organisms that produce a starch hydrolyzing enzyme will display a clear zone around the area of growth while other areas of the agar containing starch will turn blue-black.
- Starch agar is inoculated and incubated at 37 °C
- After 48 hours the surface of the agar is flooded with potassium/potassium iodide solution
To prepare media, add 25 g of starch agar to a liter of distilled or de-ionized water. Follow instructions. The media will not be sterile until autoclaved prior to use.