Hematology Stain Packs, Azer Scientific
Supplier: AZER SCIENTIFIC INC
ES901 ES900
95057-486CS 369.24 USD
95057-486 95057-484
Hematology Stain Packs, Azer Scientific
Stains Hematology Stains
Hematology stain packs are designed for use in Hema-Tek™ I, II, 1000, and 2000 model bench top automated stainers.
  • Available with modified Wright or modified Wright-Giemsa stain formula
Hematology stain packs come in a convenient carton that contains a stain, buffer, and rinse solution. Each stain pack contains enough solution to complete 1000 peripheral blood smears.