DiO/DPA Membrane potential detection kit stain for histology
Supplier: Biotium
30037
89138-972EA 347.66 USD
89138-972
DiO/DPA Membrane potential detection kit stain for histology
DiO/DPA Membrane potential detection kit
The DiO/DPA system detects cytoplasmic membrane potential changes using the principle of fluorescence resonance energy transfer (FRET), where DiO is a “stationary” FRET donor while DPA acts as a potentiometric FRET acceptor. The DiO/DPA system has been reported to produce a fluorescence signal change of >56% in HEK-293 cells and >25% in neuronal cultures and brain slices per 100 mV membrane potential change.
  • Colors: Green
  • Excitation/Emission: 484/501 nm
The DiO/DPA membrane potential detection kit comprises a vial of the green fluorescent membrane dye DiOC16(3) dissolved in DMSO (1 mL at 2 mM) and a vial of the anionic membrane quencher dye DPA (dipicrylamine) dissolved in DMSO (1 mL at 20 mM).
The DiOC16(3) in this kit is an improved version over the DiOC16(3) from other suppliers; our DiOC16(3) comprises gluconate as the counter ion, which makes the dye more water soluble, thus facilitating cell staining. DPA is traditionally a hazardous material due to its explosive nature. To improve its safety during shipping and handling, we have made it available as a diluted solution or as a solid in a smaller and more convenient packaging unit (See catalog no. 60037).