Tool Holder, Triton Products
Supplier: Triton Products
72100
76250-164PK 18.45 USD
76250-164
Tool Holder, Triton Products
Tools Storage
Tool holders accommodate a variety of tools keeping them organized and easily accessible. For the craftsman, no more digging through the tool box to find the hammer, chisel, screwdrivers or wrenches.
  • Hooks mount vertically and horizontally in four distinct positions
  • Double locking system locks the hook in place with a locking tab that mounts to the pegboard from the front
  • No need to remove the board for installation or repositioning of DuraHooks
  • Use on any 1/8" or 1/4" pegboard or DuraBoard
Ready to make that necklace for Aunt Jean, have the pliers, shears and cutters at your fingertips. What about the scrapbook you promised your sister for the new baby – hang a variety of punches and scissors in one place.