ADC® Adsafe Pocket Resuscitator
Supplier: AMERICAN DIAGNOSTIC CORP CP
4053M 4053V 4053
470315-614CS 1455.65 USD
470315-614 470315-616 470315-776
ADC® Adsafe Pocket Resuscitator
First Aid Supplies CPR Products
Functionality and portability in a single package.
  • Disposable valve to avoid cross contamination
  • Elastic green strap to keep mask in place on patient
  • Not made with natural rubber latex
  • Pocket storage case
Simplify CPR with this convenient and easily accessible mask. This CPR pocket resuscitator is designed for mouth to mask ventilation of a non-breathing adult, child, or infant. It is also used as a barrier that will direct expired air from the patient away from the user. Ergonomically shaped mask with soft-air cushion for leak-free performance, easy grasp, and patient comfort. The mask is sized for an adult, and includes an elastic band to help keep it in place. It can be used with children or infants, by inverting the mask. A high efficiency and low resistance one-way valve minimizes the possibility of cross-contamination. When assistance arrives, the 15 mm Ext.Ø connector can be attached to any standard ventilation and oxygen equipment.