Hydrion® Lo Ion pH Test Kit, Micro Essential Laboratory
Supplier: Micro Essential Laboratory
LI-5000
60720-009CS 177.58 USD
60720-009
Hydrion® Lo Ion pH Test Kit, Micro Essential Laboratory
Environmental Test Kits
The Hydrion wide range Lo ion pH test kit is intended to test weakly buffered solutions and will give accurate pH readings at each full pH unit from pH 1.0-11.0. Estimates can be made to 0.5 ph units.
The color chart has pH matches at [1.0][2.0][3.0][4.0][5.0][6.0][7.0][8.0][9.0][10.0][11.0].
Delivery information: Each kit contains a 15-foot roll of test paper, matching color chart and one(1) capped 4 ml. sample tube, packed in a polypropylene protective case.