Latex Powder-Free, Fully Textured Disposable Gloves, Tronex®
Supplier: Tronex
3667-10 3667-20 3667-30 3667-35
76208-548BX 12.38 USD
76208-548 76208-550 76208-552 76208-554
Latex Powder-Free, Fully Textured Disposable Gloves, Tronex®
Gloves
Latex powder-free gloves are food grade safe, fully textured, and they offer superior comfort and tactile sensitivity.
  • Advanced material formulation offering optimal comfort, performance, and durability
  • Powder-free (100%)
  • Fully textured for sustained grip during use
  • Superior elasticity, tactile sensitivity, and tear resistance
  • Ambidextrous with beaded cuffs
Advanced natural rubber latex material formulation is designed for optimal comfort, performance, and durability in food handling, food preparation, and light industrial applications.
Packaging: Gloves are packaged 100 per box, 10 boxes per case.