Metal-Out Static Shielding Bags, 1500 Series, SCS, Desco Industries
Supplier: Desco
150610 150Z810
89024-258PK 22.6 USD
89024-258 89024-304
Metal-Out Static Shielding Bags, 1500 Series, SCS, Desco Industries
Bags Storage Bags
Transparent, metal-out static shielding bags are designed to provide static safe packaging for the shipment and/or storage of ESD sensitive items, including outside ESD protected areas. Bags are printed with an ESD protective symbol and a lot code for traceability. Open Top and ZipTop styles are available. The bags are heat sealable.
  • Exterior Surface Rs 1×104 to < 1×108 ohms per ANSI/ESD STM11.11; Metal-Out lower exterior resistance retains less electrostatic charge than Metal-In bags
  • Metal-Out Shielding Laminate 0.003" thick (3 mil); protects ESD sensitive contents from electrostatic fields and electrostatic discharges (ESD) outside the ESD Protected Area
  • <10 nJ Discharge Shielding Energy Penetration Test per ANSI/ESD STM11.31
  • Dissipative Inner Surfaces Rs 1×10⁴ to < 1×10¹¹ ohms per ANSI/ESD STM11.11 meets Static Control Bag ANSI/ESD S11.4 Level 3
  • ElectroStatic Charges removed when placed on a grounded surface or handled by a grounded operator
SCS Metal-Out Static Shielding Bags 1500 are manufactured from a top coated metallized polyester and polyethylene laminate. The proprietary top coating is designed to protect the metal layer and quickly discharge any electrical field. Polyester dielectric, in concert with the metal layer, provides shielding for the bag contents from Electrostatic Discharge (ESD) and helps minimize the penetration of an electrical field.
Certifications: Lead-free RoHS 2, REACH and Conflict Minerals compliant.
Meets ANSI/ESD S20.20 and ANSI/ESD S541 requirements for ESD shielding packaging inside and outside an ESD Protected Area (EPA).