Hydrocortisone ≥99% (by HPLC), white solid (chromatographic standard), Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

386698-25MG
80058-206EA 172.83 USD
80058-206
Hydrocortisone ≥99% (by HPLC), white solid (chromatographic standard), Sigma-Aldrich®
Hydrocortisone

Hydrocortisone, is a corticosteroid with anti-inflammatory properties.

Formula: C₂₁H₃₀O₅
MW: 362.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011654
CAS Number: 50-23-7

Order Now

Specification Test Results

Purity ≥99% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR