Exel International® Luer Lock Holder with Pre-attached Multi-Sample Adapter, Air-Tite Products
Supplier: Air-Tite
EBCA
76290-394BX 52.66 USD
76290-394
Exel International® Luer Lock Holder with Pre-attached Multi-Sample Adapter, Air-Tite Products
Blood Collection Tube Holders
EXEL International® Luer lock holder with pre-attached multi-sample adapter are easy to use quality devices at a great value.
  • Color-coded threads
These holders provide the security of a threaded locking Luer connection. They have blue color coded threads to provide easy contrast between the holder and the Luer lock connector, providing a needle stick-free option for blood collection. Simply connect the device to an IV port using the threaded, locking connection point. The device can be used in any port compatible with female Luer connectors, including Luer activated ports found on some Foley catheters.
Certifications: Meets ISO Standards, CE registered.
Packaging: Sterile and individually wrapped.