Water USP for injection (WFI), sterile

Supplier: BAXTER IV SOLUTIONS

Synonyms: Dihydrogen oxide

2B0309 2B0309
68000-955CS 481.86 USD
68000-955 COL68000955HY
Water USP for injection (WFI), sterile
Water

Sterile Water for Injection USP.


  • pH 5.5


Packaging: Packed in a VIAFLEX plastic container. Pharmacy bulk package.

Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR