Medical-Grade Mobile Power Retrofit Kit, Tripp Lite
Supplier: Tripp Lite
HCRK-36
10144-484EA 1848.35 USD
10144-484
Medical-Grade Mobile Power Retrofit Kit, Tripp Lite
Power Supplies
The medical-grade mobile power retrofit kit provides UL 60601-1 compliance (a required patient shock-prevention measure) for all connected mobile electronic devices. The mobile power Rretrofit kit includes everything needed to add AC power to medical carts. The kit's power supply features an isolation transformer that reduces the cumulative shock potential of connected equipment, helping to ensure UL 60601-1 compliance.
  • Provides UL 60601-1 compliance for connected mobile equipment (used in every sector of healthcare facilities, both inside and outside of patient-care areas)
  • Adds reliable mobile AC power source to medical carts
  • Saves money by upgrading new or existing non-powered carts for a fraction of the cost of buying new powered models
  • Simultaneously powers connected equipment while recharging batteries
The mobile power retrofit kit also includes a remote user interface (RUI), mounting hardware and standard cabling. Medical grade mobile power retrofit kit is fully compatible with most popular carts by AFC, Anthro, Datalux, GCX, Hergo, Humanscale, Jaco, Knurr, Stinger and more.
Ordering information: Includes everything needed to retrofit a cart: mobile power supply, 36 A hour user replaceable battery module, power alert data-saving software, remote user interface (RUI) and mounting hardware kit. Two year power module and six month battery module limited warranty.