Brilliant cresyl blue solution biological stain, for microscopy, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: BCBBrilliant Blue CBrilliant cresyl blue zinc chloride double salt1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1)CI 51010

1.01384.0100 1.01384.0100
EM1.01384.0100CS 353.83 USD
CA1.01384.0100 EM1.01384.0100
Brilliant cresyl blue solution biological stain, for microscopy, Sigma-Aldrich®
Brilliant Cresyl Blue

The Brilliant cresyl blue solution is used in hematology as reagent for the staining of blood for the subsequent manual counting of blood cells. The regenerative capacity of erythrocytes can be monitored with the ratio of reticulocytes. In addition, the determination of the Substantia granulo-filamentosa (ribonucleoproteins) is possible with fresh, unfixed young erythrocytes (in a supra-vitral staining).


The Brilliant cresyl blue solution is an IVD & CE product and 100 ml of the solution are capable of staining up to 5000 slides.

Formula: C₃₄H₄₂Cl₄N₈O₂Zn
MW: 801.96 g/mol
Density: 1.01 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148901
CAS Number: 81029-05-2
IVDLogo

Order Now

Specification Test Results

Application test (Reticulocyte staining) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR