Lowenstein Jensen Medium Base, Granulated (L.J. Medium, Granulated), HiMedia Laboratories
Supplier: HiMedia
GM162-2.5KG GM162-500G GM162-5KG
75803-392EA 670.68 USD
75803-392 75803-394 75803-396
Lowenstein Jensen Medium Base, Granulated (L.J. Medium, Granulated), HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
This medium supports the growth of a wide variety of Mycobacteria and can also be used for niacin testing. This medium can be employed to identify saprophytes at room temperature (25°C) and also to identify presence or absence of pigmentation by photochromogenes and scotochromogenes at 35°C alternately in light and dark as per the type of organism.