p-Xylene-2-sulfonic acid hydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2,5-Dimethylbenzenesulphonic acid, 2,5-Dimethylbenzenesulfonic acid

X3036-100GM X3036-25GM
75873-570EA 328.35 USD
75873-570 75873-572
p-Xylene-2-sulfonic acid hydrate
p-Xylene-2-sulfonic acid hydrate
Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064858
CAS Number: 609-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR