L(-)-Tyrosine

Supplier: Spectrum Chemical Mfg. Corp.

TY105-100GM TY105-1KG TY105-5KG
75873-534EA 93.51 USD
75873-534 75873-536 75873-538
L(-)-Tyrosine
L(-)-Tyrosine

L-Tyrosine - Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR