Tyloxapol USP

Supplier: Spectrum Chemical Mfg. Corp.

T1708-100GMBL T1708-1GM T1708-25GM T1708-5GM
76014-972EA 3165.05 USD
76014-972 76014-974 76014-976 76014-978
Tyloxapol USP
Tyloxapol

All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Density: 1.100
Storage Temperature: Ambient
MDL Number: MFCD00149002
CAS Number: 25301-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR