Taurocholic acid sodium salt (natural)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Cholaic acid sodium saltSodium taurocholateSodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonate (1:1)Taurocholic acid sodiumSodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate (1:1)

SO208-25GM SO208-5GM SO208-500GMBL SO208-100GM
75873-356EA 1728.22 USD
75873-356 75873-358 75873-362 75873-360
Taurocholic acid sodium salt (natural)
Taurocholic acid sodium salt

Sodium Taurocholate, NaturalUngraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₆H₄₄NNaO₇S
MW: 537.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013050
CAS Number: 145-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR