L(+)-Serine

Supplier: Spectrum Chemical Mfg. Corp.

SE125-1KG SE125-25GM
75873-350EA 133.78 USD
75873-350 75873-352 75873-354
L(+)-Serine
L(+)-Serine

L-Serine, also known as 2-Amino-3-hydroxypropanoic acid, is one of the proteinogenic amino acids and classified as a polar amino acid.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR