Quinaldine red

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-(2-(4-Dimethylamino)styryl)-1-ethylquinolinium iodide, 2-(4-(Dimethylamino)styryl)-1-ethylquinolin-1-ium iodide

Q2024-25GM Q2024-5GM
75873-288EA 245.55 USD
75873-288 75873-302
Quinaldine red
Quinaldine red

Quinaldine Red, also known as 1-Ethyl-2-p-dimethylaminostyrylquinoline iodide, can be used as a bleaching agent and as fluorescent probe.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₁H₂₃IN₂
Storage Temperature: Ambient
MDL Number: MFCD00011968
CAS Number: 117-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR