n-Pentadecane
Supplier: Spectrum Chemical Mfg. Corp.
Pentadecane, also known as n-Pentadecane, is an alkane hydrocarbon.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
H₃C(CH₂)₁₃CH₃ Density: 0.769 Storage Temperature: Ambient |
MDL Number:
MFCD00008990 CAS Number: 629-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...