1-Phenyl-1,3-butandione

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-Phenylbutane-1,3-dione, 1-Benzoylacetone, Benzoylacetone, 1-Phenyl-1,3-butanedione

P1670-100GM P1670-25GM
75873-182EA 310.7 USD
75873-182 75873-184
1-Phenyl-1,3-butandione
1-Phenyl-1,3-butandione
Formula: C₆H₅C(O)CH₂C(O)CH₃
MW: 162.19 g/mol
Boiling Pt: 262
Storage Temperature: Ambient
MDL Number: MFCD00008786
CAS Number: 93-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR