Capsicum Oleoresin

Supplier: Spectrum Chemical Mfg. Corp.

O1030-100GM O1030-25GM O1030-500GM O1030-12KGBL
75873-114EA 214.1 USD
75873-114 75873-116 75873-118 76014-708
Capsicum Oleoresin
Capsicum Oleoresin

Capsicum Oleoresin, also known as pepper spray, is a lachrymatory agent used in policing.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Refrigerator
MDL Number: MFCD00197745
CAS Number: 8023-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR