4-Nitrobenzyl alcohol

Supplier: Spectrum Chemical Mfg. Corp.

N2205-100GM N2205-25GM
75873-092EA 253.7 USD
75873-092 75873-094
4-Nitrobenzyl alcohol
4-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00007376
CAS Number: 619-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR