Naloxone hydrochloride dihydrate USP

Synonyms: 17-Allyl-6-deoxy-7,8-dihydro-14-hydroxy-6-oxo-17-normorphine hydrochloride

N1231-250MG N1231-25GM N1231-1GM N1231-5GM N1231-300GMBL
76014-676EA 327.42 USD
76014-676 76014-678 76014-674 76014-680 76014-682
Naloxone hydrochloride dihydrate USP
Naloxone hydrochloride dihydrate

Naloxone Hydrochloride, Dihydrate, USP is an opioid antagonist used for the complete or partial reversal of opioid overdose. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₉H₂₁NO₄·HCl·2H₂O
MW: 399.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150901
CAS Number: 51481-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR