Minoxidil sulfate

Supplier: Spectrum Chemical Mfg. Corp.

M3182-1KGBL M3182-25MG M3182-5MG
75873-026EA 4952.15 USD
75873-026 75873-028 75873-030
Minoxidil sulfate
Minoxidil sulfate
Formula: C₉H₁₅N₅O₄S
Storage Temperature: Freezer
MDL Number: MFCD06248907
CAS Number: 83701-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR