4-Methylphthalic acid

Supplier: Spectrum Chemical Mfg. Corp.

M1444-100GM M1444-25GM
75872-900EA 279.5 USD
75872-900 75872-902
4-Methylphthalic acid
4-Methylphthalic acid
Formula: C₉H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD00041946
CAS Number: 4316-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR