Isobutamben (isobutyl 4-aminobenzoate)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Aminobenzoic acid isobutyl ester

I1236-100GM I1236-25GM
75872-728EA 341.9 USD
75872-728 75872-730
Isobutamben (isobutyl 4-aminobenzoate)
Isobutamben (Isobutyl 4-aminobenzoate)
Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00017110
CAS Number: 94-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR