Hematoxylin
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol
, Hydroxybrasilin
, NSC 2785
, DcH-48
, Hydroxybrazilin
, (+)-Hematoxylin
, Natural Black I
HE115-1KGBL
HE115-5GM
HE115-10GM
HE115-25GM
HE115-100GM
75872-710EA
223.38
USD
75872-710
75872-708
75872-716
75872-714
75872-712
Hematoxylin
Hematoxylin
Hematoxylin is a a substance that is extracted from the heartwood of logwood tree and along with eosin stain is one of the most widely used stains in histology. It is a permanent stain as opposed to other stains that are only temporary in nature.
Formula:
C₁₆H₁₄O₆ MW: 302.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...