4-Benzylphenol

Supplier: Spectrum Chemical Mfg. Corp.

H2038-100GM H2038-25GM
75873-666EA 426 USD
75873-666 75873-668
4-Benzylphenol
4-Benzylphenol
Formula: C₁₃H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00002384
CAS Number: 101-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR