3-Hydroxydiphenylamine

Supplier: Spectrum Chemical Mfg. Corp.

H1496-100GM H1496-25GM
75872-626EA 279.5 USD
75872-626 75872-628
3-Hydroxydiphenylamine
3-Hydroxydiphenylamine
Formula: C₁₂H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00002262
CAS Number: 101-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR