L(+)-Glutamic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: L-Glutamic acid(S)-(+)-1-Aminopropane-1,3-dicarboxylic acidL(+)-α-Glutamic acidH-Glu-OH(S)-(+)-Glutamic acid(S)-(+)-α-Glutamic acidL(+)-1-Aminopropane-1,3-dicarboxylic acid(S)-(+)-2-Aminopentanedioic acidL(+)-2-Aminopentanedioic acid

GL125-25KGBL GL125-1KG GL125-100GM
75872-584EA 139.88 USD
75872-584 75872-582 76014-516
L(+)-Glutamic acid
L(+)-Glutamic acid

L-Glutamic Acid is also known as glutamic acid. It is used in the food industry most usually as a flavor with an umami taste.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
https://us.vwr.com/stibo/web/std.lang.all/98/27/2299827.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR