Green tea

Supplier: Spectrum Chemical Mfg. Corp.

G1283-2.5KG G1283-500GM G1283-12KG
75872-526EA 238.15 USD
75872-526 75872-528 76014-504
Green tea
Green tea

Green Tea, Powder is high in antioxidants.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 84650-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR