Furfuryl alcohol

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Furfurol

F1100-20LT F1100-1LT F1100-2.5LTPV F1100-250ML F1100-200LTBL
76014-456EA 1059.78 USD
76014-456 76014-454 75872-506 75873-656 75872-504
Furfuryl alcohol
Furfuryl alcohol

Furfuryl Alcohol, also known as 2-furylmethanol, is often used in the production of resins for bonding foundry sands for the production of molds and cores.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₆O₂
MW: 98.1 g/mol
Density: 1.135
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
UN: 2874
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR