EDTA (ethylenediamine tetraacetic acid)
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, Edetic Acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, EDTA, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
(Ethylenedinitrilo)tetraacetic Acid, Powder is generally abbreviated as EDTA, a colorless, water soluble, aminopolycarboxylic acid and solid. It's used for its chelating properties and as a ligand. It is used by many industries for application ranging from sequestering agent to chelation therapy. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for consumption.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₁₆N₂O₈ MW: 292.25 g/mol Boiling Pt: 614 Melting Pt: 220 Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 Merck Index: 13,03546 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...