Ethyl glycidyl ether

Supplier: Spectrum Chemical Mfg. Corp.

E1466-100ML E1466-25ML
75872-406EA 341.48 USD
75872-406 75872-408
Ethyl glycidyl ether
Ethyl glycidyl ether
Formula: C₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00085264
CAS Number: 4016-11-9
UN: 2752
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR