1,2-Dioleoyl-sn-glycero-3-phosphocholine

Supplier: Spectrum Chemical Mfg. Corp.

D3953-1GM D3953-500MG
75872-382EA 702.85 USD
75872-382 75872-384
1,2-Dioleoyl-sn-glycero-3-phosphocholine
1,2-Dioleoyl-sn-glycero-3-phosphocholine
Formula: C₄₄H₈₄NO₈P
Storage Temperature: Low Temperature Freezer (–40°C)
MDL Number: MFCD00135191
CAS Number: 4235-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR