2,5-Dimethylpyrazine
Supplier: Spectrum Chemical Mfg. Corp.
D2984-100ML
D2984-25ML
75872-374EA
381.25
USD
75872-374
75872-376
2,5-Dimethylpyrazine
2,5-Dimethylpyrazine
2,5-Dimethylpyrazine is a laboratory chemical, also known as 2,5-Dimethyl-p-diazine.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₈N₂ Density: 0.990 Storage Temperature: Ambient |
MDL Number:
MFCD00006147 CAS Number: 123-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...