3,4-Diaminobenzophenone

Supplier: Spectrum Chemical Mfg. Corp.

D2105-100GM D2105-25GM
75872-338EA 336.45 USD
75872-338 75872-340
3,4-Diaminobenzophenone
3,4-Diaminobenzophenone
Formula: C₁₃H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00007727
CAS Number: 39070-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR