3-Chloropyridine

Supplier: Spectrum Chemical Mfg. Corp.

C2919-100GM C2919-25GM
75872-166EA 274.05 USD
75872-166 75872-168
3-Chloropyridine
3-Chloropyridine
Formula: C₅H₄ClN
Density: 1.194
Storage Temperature: Ambient
MDL Number: MFCD00006375
CAS Number: 626-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR