4-Chlorobenzonitrile

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: p-Chlorobenzonitrile

C2250-100GM C2250-25GM
75872-152EA 251 USD
75872-152 75872-154
4-Chlorobenzonitrile
4-Chlorobenzonitrile
Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 223
Storage Temperature: Ambient
MDL Number: MFCD00001813
CAS Number: 623-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR