[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex
B3186-1GM
B3186-25GM
B3186-5GM
75872-068EA
90.25
USD
75872-068
75872-070
75872-072
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
[1,1'-Bis(diphenylphosphino)ferrocene]palladium (II) chloride dichloromethane complex
Formula:
C₃₅H₃₀Cl₄FeP₂Pd MW: 816.64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00792899 CAS Number: 95464-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...