Benzoin methyl ether

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone

B2523-25GM B2523-100GM
75871-834EA 87.5 USD
75871-834 75872-058
Benzoin methyl ether
Benzoin methyl ether
Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008492
CAS Number: 3524-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR