α-Benzoinoxime

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Hydroxy-1,2-diphenylethanone oxime, α-Benzoin oxime

B2105-100GM B2105-25GM
75872-034EA 219.8 USD
75872-034 75872-036
α-Benzoinoxime
α-Benzoinoxime
Formula: C₁₄H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00004501
CAS Number: 441-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR