L-Arginine hydrochloride
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(S)-(+)-Arginine hydrochloride
, L-Arg
, L-Arginine, hydrochloride
, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride
, L(+)-Arginine hydrochloride
, L(+)-Arginine monohydrochloride
, (S)-(+)-Arginine HCl
, H-Arg-OH·HCl
, H-L-Arg-OH.HCl
, H-Arg-OH.HCl
L-Arginine Hydrochloride is an alpha-amino acid that is a semi-essential amino acid. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use and typically are not suitable for consumption.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₁₅ClN₄O₂ MW: 210.66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064550 CAS Number: 1119-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...