4-(Aminomethyl)benzoic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

A2588-100GM A2588-25GM
75871-802EA 324.25 USD
75871-802 75871-844
4-(Aminomethyl)benzoic acid
4-(Aminomethyl)benzoic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010203
CAS Number: 56-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR