Hexyl salicylate

Supplier: Pfaltz & Bauer
Warning

Synonyms: Hexyl 2-hydroxybenzoateN-Hexyl salicylate

H06585-100G
101187-230EA 420.38 USD
101187-230
Hexyl salicylate
Hexyl salicylate
Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Boiling Pt: 290 °C
Density: 1,04 g/cm³
Flash Pt: 122 °C
MDL Number: MFCD00036487
EINECS: 228-408-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR